- Genetic and functional analysis of two missense
Genetic and functional analysis of two missense
Oncotarget (2016-07-11)
Shiguo Liu, Wenhui Zhang, Liqin Zhang, Hui Zou, Kunna Lu, Qiang Li, Hongfei Xia, Shengli Yan, Xu Ma
PMID29435108
ABSTRACT
Mutations in the dual oxidase 2 gene (