- Neuroprotective effect of the active components of three Chinese herbs on brain iron load in a mouse model of Alzheimer's disease.
Neuroprotective effect of the active components of three Chinese herbs on brain iron load in a mouse model of Alzheimer's disease.
Experimental and therapeutic medicine (2015-03-18)
Xian-Hui Dong, Wei-Juan Gao, Wei-Na Kong, Hong-Lin Xie, Yan Peng, Tie-Mei Shao, Wen-Guo Yu, Xi-Qing Chai
PMID25780429
ABSTRACT
Alzheimer's disease (AD) is a neurodegenerative brain disorder and the most common cause of dementia. New treatments for AD are required due to its increasing prevalence in aging populations. The present study evaluated the effects of the active components of