Merck
  • Adenosine receptor expression in an experimental animal model of myocardial infarction with preserved left ventricular ejection fraction.
Adenosine receptor expression in an experimental animal model of myocardial infarction with preserved left ventricular ejection fraction.
Heart and vessels (2013-07-12)
Manuela Cabiati, Alessandro Martino, Letizia Mattii, Chiara Caselli, Tommaso Prescimone, Vincenzo Lionetti, Maria-Aurora Morales, Silvia Del Ry
ABSTRACT
Adenosine, a purine nucleoside and a "retaliatory metabolite" in ischemia, is ubiquitous in the body and increases 100-fold during ischemia. Its biological actions are mediated by four adenosine receptors (ARs): A(1), A(2A), A(2B) and A(3). The aim of this study was to determine possible myocardial alterations in AR expression in an experimental animal model of myocardial infarction (MI) with a preserved left ventricular (LV) ejection fraction. LV tissue was collected from sexually mature male farm pigs with MI (n = 6) induced by permanent surgical ligation of the left anterior descending coronary artery and from five healthy pigs (C). mRNA expression of A(1)R, A(2A)R, A(2B)R, A(3)R and TNF-α was determined by real-time PCR in tissue collected from border (BZ) and remote zones (RZ) of the infarcted area and from LV of C. BZ, RZ and samples of C were stained immunohistochemically to investigate A(3)R immunoreaction. After 4 weeks a different regulation of ARs was observed. A(1)R mRNA expression was significantly lower in the infarct regions than in controls (C = 0.75 ± 0.2, BZ = 0.05 ± 0.2, RZ = 0.07 ± 0.02 p = 0.0025, p = 0.0016, C vs. BZ and RZ, respectively). Conversely A(3)R was higher in infarct areas (C = 0.94 ± 0.2, BZ = 2.85 ± 0.5, RZ = 3.48 ± 1.0, p = 0.048 C vs. RZ). No significant differences were observed for A(2A)R (C = 1.58 ± 0.6, BZ = 0.42 ± 0.1, RZ = 1.37 ± 0.6) and A(2B)R (C = 1.66 ± 0.2, BZ = 1.54 ± 0.5, RZ = 1.25 ± 0.4). A(3)R expression was confirmed by immunohistochemical analysis and was principally localized in cardiomyocytes. TNF-α mRNA results were: C 0.41 ± 0.25; BZ 1.60 ± 0.19; RZ 0.17 ± 0.04. The balance between A(1)R and A(3)R as well as between A(2A)R and A(2B)R was consistent with adaptative retaliatory anti-ischemic adenosinergic changes in the infarcted heart with preserved LV function.
Product Description
Supelco
Residual Solvent - Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, puriss. p.a., reag. ISO, reag. Ph. Eur., 99.0-99.4% (GC)
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, amylene stabilized
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains amylenes as stabilizer
Supelco
Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Supelco
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Supelco
Chloroform, analytical standard
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%