- ROR1 expression correlated with poor clinical outcome in human ovarian cancer.
ROR1 expression correlated with poor clinical outcome in human ovarian cancer.
The receptor-tyrosine-kinase-like orphan receptor 1 (ROR1) is a transmembrane protein belongs to receptor tyrosine kinase (RTK) family. This study aimed to examine the expression of ROR1 in human ovarian cancer and investigate the relationship between its expression and the prognosis of ovarian cancer patients. In this present study, one-step quantitative reverse transcription-polymerase chain reaction (15 ovarian cancer samples of high FIGO stage, 15 ovarian cancer samples of low FIGO stage and nine normal ovary tissue samples) and immunohistochemistry by tissue microarrays (100 ovarian cancer samples and 50 normal ovary samples) were performed to characterize expression of the ROR1 gene in ovarian cancer. Kaplan-Meier survival and Cox regression analyses were executed to evaluate the prognosis of ovarian cancer. The results of qPCR and IHC analysis showed that the expression of ROR1 in ovarian cancer was significantly higher than that in normal ovary tissues (all p < 0.05). Survival analysis showed that ROR1 protein expression was one of the independent prognostic factors for disease-free survival and overall survival (both p < 0.05). The data suggest that ROR1 expression is correlated with malignant attributes of ovarian cancer and it may serve as a novel prognostic marker in ovarian cancer.