Merck
  • Control of NF-kB activity in human melanoma by bromodomain and extra-terminal protein inhibitor I-BET151.
Control of NF-kB activity in human melanoma by bromodomain and extra-terminal protein inhibitor I-BET151.
Pigment cell & melanoma research (2014-06-14)
Stuart J Gallagher, Branka Mijatov, Dilini Gunatilake, Kavitha Gowrishankar, Jessamy Tiffen, Wilmott James, Lei Jin, Gulietta Pupo, Carleen Cullinane, Grant A McArthur, Peter J Tummino, Helen Rizos, Peter Hersey
ABSTRACT
The transcription factor NF-kappaB (NF-kB) is a key regulator of cytokine and chemokine production in melanoma and is responsible for symptoms such as anorexia, fatigue, and weight loss. In addition, NF-kB is believed to contribute to progression of the disease by upregulation of cell cycle and anti-apoptotic genes and to contribute to resistance against targeted therapies and immunotherapy. In this study, we have examined the ability of the bromodomain and extra-terminal (BET) protein inhibitor I-BET151 to inhibit NF-kB in melanoma cells. We show that I-BET151 is a potent, selective inhibitor of a number of NF-kB target genes involved in induction of inflammation and cell cycle regulation and downregulates production of cytokines such as IL-6 and IL-8. SiRNA studies indicate that BRD2 is the main BET protein involved in regulation of NF-kB and that I-BET151 caused transcriptional downregulation of the NF-kB subunit p105/p50. These results suggest that BET inhibitors may have an important role in treatment of melanoma where activation of NF-kB may have a key pathogenic role.
Sigma-Aldrich
Rat IL-6 ELISA Kit, for serum, plasma and cell culture supernatants
Sigma-Aldrich
Rat Interleukin-6 ELISA Kit, for for cell lysate and tissue lysate
Sigma-Aldrich
Mouse IL-6 R ELISA Kit, for serum, plasma and cell culture supernatant
Sigma-Aldrich
Mouse IL-6 ELISA Kit, for cell and tissue lysates