Merck
  • Functional integrity of the retrosplenial cortex is essential for rapid consolidation and recall of fear memory.
Functional integrity of the retrosplenial cortex is essential for rapid consolidation and recall of fear memory.
Learning & memory (Cold Spring Harbor, N.Y.) (2013-03-19)
Cynthia Katche, Guido Dorman, Leandro Slipczuk, Martín Cammarota, Jorge H Medina
ABSTRACT
Memory storage is a temporally graded process involving different phases and different structures in the mammalian brain. Cortical plasticity is essential to store stable memories, but little is known regarding its involvement in memory processing. Here we show that fear memory consolidation requires early post-training macromolecular synthesis in the anterior part of the retrosplenial cortex (aRSC), and that reversible pharmacological inactivation of this cortical region impairs recall of recent as well as of remote memories. These results challenge the generally accepted idea that neocortical areas are slow encoding systems that participate in the retrieval of remote memories only.
Sigma-Aldrich
Anisomycin from Streptomyces griseolus, ≥98% (HPLC), solid
Sigma-Aldrich
Anisomycin, Ready Made Solution from Streptomyces griseolus, 10 mg/mL in DMSO