Merck
  • Newly identified genetic variations in common Escherichia coli MG1655 stock cultures.
Newly identified genetic variations in common Escherichia coli MG1655 stock cultures.
Journal of bacteriology (2011-11-15)
Peter L Freddolino, Sasan Amini, Saeed Tavazoie
ABSTRACT
We have recently identified seven mutations in commonly used stocks of the sequenced Escherichia coli strain MG1655 which do not appear in the reference sequence. The mutations are likely to cause loss of function of the glpR and crl genes, which may have serious implications for physiological experiments using the affected strains.
Sigma-Aldrich
DL-α-Glycerol phosphate magnesium salt hydrate, ~85% (KT)