Merck
  • Online enzyme discrimination and determination of substrate enantiomers based on electrophoretically mediated microanalysis.
Online enzyme discrimination and determination of substrate enantiomers based on electrophoretically mediated microanalysis.
Analytical chemistry (2012-07-04)
Wenwen Zhao, Miaomiao Tian, Rongbin Nie, Yulin Wang, Liping Guo, Li Yang
ABSTRACT
We proposed the first application of an electrophoretically mediated microanalysis (EMMA) method for fast online discrimination and determination of substrate enantiomers, which was achieved by just one EMMA assay. Lactate dehydrogenase (LDH)-catalyzed reaction was studied to evaluate the feasibility and performance of the presented method. The L- and D-LDH chiral enzymatic reactions, which are highly stereoselective to the lactate enantiomers, were initiated successively in one capillary, and the corresponding products, nicotinamide adenine dinucleotide (NADH), were online discriminated and detected by UV absorption. Excellent linear dependence of the two NADH peak intensities on the concentration of the corresponding lactate enantiomers was obtained within a wide range of 0.1-10 mM. The limit of detection was 26 μM for D-lactate and 49 μM for L-lactate (S/N = 3). Good repeatability of online chiral discrimination was demonstrated with relative standard deviation (RSD) < 6.3% for NADH peak height and RSD < 1.5% for migration time (n = 5). K(m) values for L- and D-lactate were measured and were consistent with those of the off-line enzyme assays. The presented method was successfully applied to determine the L-/D-lactate in several yogurt and wine samples. Our study shows a new application of the EMMA method utilizing high stereoselectivity of enzymes for fast online chiral analysis.
Sigma-Aldrich
D-Lactic Dehydrogenase from Staphylococcus epidermidis, lyophilized powder, ≥80 units/mg solid
Sigma-Aldrich
D-Lactic Dehydrogenase from Lactobacillus leichmannii, lyophilized powder, 150-500 units/mg protein
Sigma-Aldrich
D-Lactic Dehydrogenase from Lactobacillus leichmannii, ammonium sulfate suspension, ≥250 units/mg protein (biuret)