Merck
  • STING is an essential regulator of heart inflammation and fibrosis in mice with pathological cardiac hypertrophy via endoplasmic reticulum (ER) stress.
STING is an essential regulator of heart inflammation and fibrosis in mice with pathological cardiac hypertrophy via endoplasmic reticulum (ER) stress.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie (2020-02-29)
Yan Zhang, Wenzhong Chen, Yan Wang
ABSTRACT
Pathological cardiac hypertrophy is characterized by myocyte enlargement and cardiac dysfunction. However, the pathogenesis for this disease is still poorly understood. Stimulator of interferon genes (STING) could meditate inflammation and immune response in various kinds of diseases. In this work, we demonstrated that STING was critical for pressure overload-induced cardiac hypertrophy. Results showed that STING expression was up-regulated in human and mouse hypertrophic hearts. STING knockout attenuated cardiac hypertrophy induced by aortic banding (AB). The effects of STING deficiency on the improvement of cardiac hypertrophy and dysfunction were associated with the restrained macrophage infiltration, inflammatory response and fibrosis. Moreover, ER stress was detected in hearts of AB-operated mice, as evidenced by the increased expression of phospho-protein kinase RNA-like endoplasmic reticulum kinase (PERK), phospho-eukaryotic initiation factor 2 alpha (eIF2α) and phospho-inositol-requiring kinase (IRE)-1α. Importantly, these proteins were restrained in mice with STING knockout after AB surgery. What's more, angiotensin II (Ang II)-induced STING could be accelerated by ER stress activator, while being markedly abolished by the ER stress inhibitor. We then found that whether co-treated with or without transforming growth factor-beta 1 (TGF-β1), cardiac fibroblasts cultured in the conditional medium (CM) from Ang II-incubated cardiomyocytes with STING knockdown exhibited significantly reduced fibrosis, as displayed by the clearly down-regulated expression of α-SMA, Collagen type I (Col I) and Collagen type III (Col III). Therefore, we defined STING as an important signal contributing to cardiac hypertrophy closely associated with ER stress.
Sigma-Aldrich
4-Phenylbutyric acid, 99%
Sigma-Aldrich
MISSION® esiRNA, targeting human EIF2AK3, AC104134.2
Sigma-Aldrich
Thapsigargin, ≥98% (HPLC), solid film