Merck
  • Illumination/Darkness-Induced Changes in Leaf Surface Potential Linked With Kinetics of Ion Fluxes.
Illumination/Darkness-Induced Changes in Leaf Surface Potential Linked With Kinetics of Ion Fluxes.
Frontiers in plant science (2019-12-04)
Jinhai Li, Yang Yue, Ziyang Wang, Qiao Zhou, Lifeng Fan, Zhiqiang Chai, Chao Song, Hongtu Dong, Shixian Yan, Xinyu Gao, Qiang Xu, Jiepeng Yao, Zhongyi Wang, Xiaodong Wang, Peichen Hou, Lan Huang
ABSTRACT
A highly reproducible plant electrical signal-light-induced bioelectrogenesis (LIB) was obtained by means of periodic illumination/darkness stimulation of broad bean (Vicia faba L.) leaves. By stimulating the same position of the same leaf with different concentrations of NaCl, we observed that the amplitude and waveform of the LIB was correlated with the intensity of stimulation. This method allowed us to link dynamic ion fluxes induced by periodic illumination/darkness to salt stress. The self-referencing ion electrode technique was used to explore the ionic mechanisms of the LIB. Fluxes of H+, Ca2+, K+, and Cl- showed periodic changes under periodic illumination/darkness before and after 50 mM NaCl stimulation. Gray relational analysis was used to analyze correlations between each of these ions and LIB. The results showed that different ions are involved in surface potential changes at different stages under periodic illumination/darkness. The gray relational grade reflected the contribution of each ion to the change in surface potential at a certain time period. The ion fluxes data obtained under periodic illumination/darkness stimulation will contribute to the future development of a dynamic model for interpretation of electrophysiological events in plant cells.
Product Description
Supelco
Potassium ionophore I - cocktail B, Selectophore
SKU
Pack Size
Availability
Price
Quantity
Supelco
Chloride ionophore I - cocktail A, Selectophore