Merck
  • Regulation of Respiration and Apoptosis by Cytochrome c Threonine 58 Phosphorylation.
Regulation of Respiration and Apoptosis by Cytochrome c Threonine 58 Phosphorylation.
Scientific reports (2019-11-05)
Junmei Wan, Hasini A Kalpage, Asmita Vaishnav, Jenney Liu, Icksoo Lee, Gargi Mahapatra, Alice A Turner, Matthew P Zurek, Qinqin Ji, Carlos T Moraes, Maurice-Andre Recanati, Lawrence I Grossman, Arthur R Salomon, Brian F P Edwards, Maik Hüttemann
ABSTRACT
Cytochrome c (Cytc) is a multifunctional protein, acting as an electron carrier in the electron transport chain (ETC), where it shuttles electrons from bc1 complex to cytochrome c oxidase (COX), and as a trigger of type II apoptosis when released from the mitochondria. We previously showed that Cytc is regulated in a highly tissue-specific manner: Cytc isolated from heart, liver, and kidney is phosphorylated on Y97, Y48, and T28, respectively. Here, we have analyzed the effect of a new Cytc phosphorylation site, threonine 58, which we mapped in rat kidney Cytc by mass spectrometry. We generated and overexpressed wild-type, phosphomimetic T58E, and two controls, T58A and T58I Cytc; the latter replacement is found in human and testis-specific Cytc. In vitro, COX activity, caspase-3 activity, and heme degradation in the presence of H2O2 were decreased with phosphomimetic Cytc compared to wild-type. Cytc-knockout cells expressing T58E or T58I Cytc showed a reduction in intact cell respiration, mitochondrial membrane potential (∆Ψm), ROS production, and apoptotic activity compared to wild-type. We propose that, under physiological conditions, Cytc is phosphorylated, which controls mitochondrial respiration and apoptosis. Under conditions of stress Cytc phosphorylations are lost leading to maximal respiration rates, ∆Ψm hyperpolarization, ROS production, and apoptosis.
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Corning®  96 Well Black Polystyrene Microplate, flat bottom clear, black polystyrene plate (Ideal for fluorescent assays), Tissue Culture (TC)-treated surface, pkg of (individually wrapped), sterile, lid