Merck
  • An in vitro and in silico investigation of human pregnane X receptor agonistic activity of poly- and perfluorinated compounds using the heuristic method-best subset and comparative similarity indices analysis.
An in vitro and in silico investigation of human pregnane X receptor agonistic activity of poly- and perfluorinated compounds using the heuristic method-best subset and comparative similarity indices analysis.
Chemosphere (2019-09-29)
Yi-Ming Zhang, Tao Wang, Xu-Shu Yang
ABSTRACT
Poly- and perfluorinated compounds (PFCs) may induce potential endocrine-disrupting hormonal effects. However, the molecular mechanism of the toxicology of PFCs remains unclear, and the insufficient information is available on the biological activities of PFCs at present. In this study, the cell-based reporter gene assays were used to determine the agonistic activity of PFCs on the human pregnane X receptor (hPXR). The heuristic method combined with best subset modeling (HM-BSM) based on Dragon descriptors and comparative similarity indices analysis (CoMSIA) were employed to build classical quantitative structure-activity relationship (QSAR) and three-dimensional QSAR models, respectively. The applicability domain (AD) of the classical QSAR model was assessed. Both the HM-BSM and CoMSIA approaches demonstrated good robustness, predictive ability, and mechanistic interpretability. The r2 and leave-one-out cross-validation squared correlated coefficient (q2LOO) values were 0.872 and 0.759 for the HM-BSM, and 0.976 and 0.751 for the CoMSIA model, respectively. The hPXR agonistic activity of the PFCs predicted by the built HM-BSM and CoMSIA agreed well with experimental activity, with root mean square error (RMSE) values of 0.0803 and 0.117, respectively, and external validation squared correlated coefficients (q2EXT) of 0.972 and 0.932, respectively. The hPXR agonistic activity of PFCs was related to their molecular polarizability, charge and atomic mass. Hydrogen bonding and hydrophobic interactions constituted the primary intermolecular forces between PFCs and the hPXR. The developed models were used to screen the PFCs with high hPXR agonistic activity.
Product Description
Supelco
Sulfluramid, PESTANAL®, analytical standard
Sigma-Aldrich
Heptadecafluoro-1-iodooctane, 98%