Merck
  • Identification of TPD52 and DNAJB1 as two novel bile biomarkers for cholangiocarcinoma by iTRAQ‑based quantitative proteomics analysis.
Identification of TPD52 and DNAJB1 as two novel bile biomarkers for cholangiocarcinoma by iTRAQ‑based quantitative proteomics analysis.
Oncology reports (2019-10-30)
Hongyue Ren, Mingxu Luo, Jinzhong Chen, Yanming Zhou, Xiumei Li, Yanyan Zhan, Dongyan Shen, Bo Chen
ABSTRACT
Cholangiocarcinoma (CCA) represents a type of epithelial cancer with a late diagnosis and poor outcome. However, the molecular mechanisms responsible for the development of CCA have not yet been fully identified. Thus, in this study, we aimed to elucidate some of these mechanisms. For this purpose, isobaric tags for relative and absolute quantification (iTRAQ) was performed to analyze the secretory proteins from the 2 CCA cell lines, TFK1 and HuCCT1, as well as from a normal biliary epithelial cell line, human intrahepatic biliary epithelial cells (HiBECs). Differentially expressed proteins (DEPs) were identified and biological process analysis was performed according to the Gene Ontology (GO) functional classification annotation and KEGG metabolic pathway map analysis. tumor protein D52 (TPD52) and DnaJ heat shock protein family (Hsp40) member B1 (DNAJB1) were validated using RT‑qPCR, western blot analysis and immunohistochemistry. In total, 778 proteins were identified as DEPs. Following validation, TPD52 and DNAJB1 were used for further analysis. The expression levels of TPD52 and DNAJB1 were elevated in CCA cell lines, tissues and bile samples, suggesting that these proteins may contribute to tumor pathogenesis. In addition, the expression levels of TPD52 and DNAJB1 were found to be closely associated with the clinical parameters and prognosis of patients with CCA. On the whole, the findings of this study indicate that TPD52 and DNAJB1 may serve as novel bile biomarkers for CCA.
Sigma-Aldrich
Anti-Mouse IgG (Fab specific)–Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-DNAJB1 antibody produced in rabbit, affinity isolated antibody