Merck
  • Wheat Bran for Colon Cancer Prevention: The Synergy between Phytochemical Alkylresorcinol C21 and Intestinal Microbial Metabolite Butyrate.
Wheat Bran for Colon Cancer Prevention: The Synergy between Phytochemical Alkylresorcinol C21 and Intestinal Microbial Metabolite Butyrate.
Journal of agricultural and food chemistry (2019-11-02)
Yantao Zhao, Lei Shi, Changling Hu, Shengmin Sang
ABSTRACT
There is convincing evidence that consuming whole grains (WGs) may decrease the risk of colorectal cancer (CRC). Wheat bran (WB) is a rich source of dietary fiber and phytochemicals with health-promoting properties. However, the active components especially the interaction between different components in WG wheat have not been fully explored. Here, we investigated whether one of the major WB phytochemicals, alkylresorcinol (AR) C21, and the major active intestinal microbial metabolite of fiber, butyrate, could synergistically suppress human colon cancer cells. Our results demonstrated for the first time that the combination of C21 and butyrate synergistically inhibited the growth of human colon cancer cells and induced apoptosis. Further mechanistic studies demonstrated that the cotreatment of C21 and butyrate induced significant up-regulations in cleaved Poly(ADP-ribose) polymerase (PARP), cleaved caspase 3, p53 upregulated modulator of apoptosis (PUMA), cytochrome C, lipid-conjugated membrane-bound form of microtubule-associated protein 1A/1B-light chain 3 (LC3-II), and C/EBP homologous protein (CHOP) expressions, indicating the synergistic anticancer effects of C21 and butyrate were associated with induction of apoptosis, autophagy, and ER stress pathways. Notably, the C21 concentrations in the large intestinal tract of mice treated with human relevant doses of C21, were from 0.86 to 1.78 μmol/g, suggesting the C21 doses used in vitro may be achievable after daily WG wheat intake. These results provide novel insights into the dietary prevention of CRC regarding the potential interaction of bioactive WG wheat phytochemicals and the microbial metabolites of fiber.
Sigma-Aldrich
DL-Glyceraldehyde, ≥90% (GC)
Sigma-Aldrich
NS3694, ≥98%, powder