Merck
  • The Molecular Identification and Antifungal Susceptibilities of Aspergillus Species Causing Otomycosis in Tochigi, Japan.
The Molecular Identification and Antifungal Susceptibilities of Aspergillus Species Causing Otomycosis in Tochigi, Japan.
Mycopathologia (2018-10-07)
Shigehiro Hagiwara, Takashi Tamura, Kazuo Satoh, Hitoshi Kamewada, Masayasu Nakano, Seiichi Shinden, Hideyo Yamaguchi, Koichi Makimura
ABSTRACT
Aspergillus species are the most common pathogenic fungi involved in otomycosis, an infection of the outer ear canal. In this study, we examined the incidence of Aspergillus infections and the antifungal susceptibilities of 30 Aspergillus species isolates from patients with otomycosis who visited Saiseikai Utsunomiya Hospital between August 2013 and July 2016. Based on the morphological test results, the strains were identified as Aspergillus niger sensu lato (20 strains), A. terreus sensu lato (7 strains), and A. fumigatus sensu lato (3 strains). In contrast, the molecular identifications based on analyzing the isolates' partial β-tubulin gene sequences revealed them to be A. niger sensu stricto (12 strains), A. tubingensis (8 strains), A. terreus sensu stricto (7 strains), and A. fumigatus sensu stricto (3 strains). The antifungal susceptibility test results indicated that strains of A. tubingensis and A. niger sensu stricto displayed lower susceptibilities to ravuconazole, compared with the other isolates. The Aspergillus strains from this study showed low minimum inhibitory concentrations toward the azole-based drugs efinaconazole, lanoconazole, and luliconazole. Therefore, these topical therapeutic agents may be effective for the treatment of otomycosis.