Merck
Unusual neurofilament composition in cerebellar unipolar brush neurons.
Journal of neurocytology (1993-12-01)
J Harris, S Moreno, G Shaw, E Mugnaini
ABSTRACT
During antibody screening on sections of rat cerebellum, we noticed a group of small neurons which exhibited unusual staining properties. They were robustly immunopositive for the high molecular weight neurofilament protein, moderately immunostained with antibodies to the low molecular weight neurofilament protein and alpha-internexin, but only faintly immunoreactive (in PAP sections) or essentially immunonegative (in immunofluorescent sections) with all members of a panel of antibodies directed against the middle molecular weight neurofilament protein. Since neurons generally react equally well with phosphate-independent, (antibodies to) low, middle and high molecular weight neurofilament protein, we conclude that middle molecular weight neurofilament protein is present in these cells in an unusually low relative amount. These cells are found in the granular layer and appear concentrated in the flocculus, ventral paraflocculus, and vermis, particularly in the ventral uvula and nodulus (lobules IXd and X). Previous studies performed by Hockfield defined a population of neurons of similar appearance and distribution using the monoclonal antibody Rat-302, which recognized an uncharacterized 160 kDa protein. We show here that the cells described by Hockfield are identical to those we have found and furthermore that the Rat-302 antibody specifically recognizes the dephosphorylated form of the lysine-serine-proline repeated sequences of high molecular weight neurofilament protein. These cells were studied by pre-embedding immunoelectron microscopy. The nucleus is deeply indented and shows little condensed chromatin. The cytoplasm contains scattered microtubules and a larger number of neurofilaments than expected in a small cell. There are numerous large dense core vesicles, an unusual organelle consisting of ringlet subunits, and relatively little granular endoplasmic reticulum. A thin axon and a single stout dendritic trunk emanate from the perikaryon. Although the cell body and the dendritic shaft may form either complex contacts with mossy fibres (resembling those previously termed en marron synapses) or simple symmetric synapses with small boutons containing pleomorphic vesicles, most of the synaptic relations are established on the shafts of brush-like branchlets that form at the tip of the dendrite and enter one or two glomeruli. Each branchlet forms an extraordinarily extensive asymmetric synapse with the mossy fibre rosette and the subsynaptic region shows a microfibrillar web connected to the postsynaptic density. In addition to other organelles, the branchlets contain numerous mitochondria and large dense core vesicles. Short, non-synaptic appendages with few cytoplasmic organelles emanated from the cell body, dendritic shaft and branchlets.(ABSTRACT TRUNCATED AT 400 WORDS)