![Triazolam European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/221/725/196f24ec-aeda-4b1d-9a42-bb4528e3c1c3/640/196f24ec-aeda-4b1d-9a42-bb4528e3c1c3.png)
Triazolam
![Triazolam European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/221/725/196f24ec-aeda-4b1d-9a42-bb4528e3c1c3/640/196f24ec-aeda-4b1d-9a42-bb4528e3c1c3.png)
Synonym(s):
8-Chloro-6-[2-chlorophenyl]-1-methyl-4H-[1,2,4]triazolo[4,3-a][1,4]benzodiazepine
Empirical Formula (Hill Notation):
C17H12Cl2N4
CAS Number:
Molecular Weight:
343.21
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing