Sulfamethazine
Synonym(s):
4,6-Dimethylsulfadiazine, 4-Amino-N-(4,6-dimethyl-2-pyrimidinyl)benzenesulfonamide, Sulfadimethyldiazine, Sulfadimidine, Sulfamethazine
Empirical Formula (Hill Notation):
C12H14N4O2S
CAS Number:
Molecular Weight:
278.33
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Activity Spectrum
Agency
Format
Quality Segment
Application
Mode of Action
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing