Trolamine
Synonym(s):
Triethanolamine, 2,2′,2′′-Nitrilotriethanol, Tris(2-hydroxyethyl)amine
Linear Formula:
(HOCH2CH2)3N
CAS Number:
Molecular Weight:
149.19
Products
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
pH Value
pK Value
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3