Skip to Content
AU
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Methanesulfonic acid concentrate
Synonym(s):
Methanesulfonic acid solution
Linear Formula:
CH
3
SO
3
H
CAS Number:
75-75-2
Molecular Weight:
96.11
EC Number:
200-898-6
Products
Product Category
organic acids (1)
Brand
Sigma-Aldrich (3)
Supelco (1)
Feature
Formula Weight
Physical Form
Quality Segment
Markush Class
Markush Group
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
55517
Methanesulfonic acid concentrate,
0.1 M CH
3
SO
3
H in water (0.1N), eluent concentrate for IC
Expand
View Pricing
471348
Methanesulfonic acid concentrate,
70 wt. % in H
2
O
Expand
View Pricing
8.14637
Methanesulfonic acid concentrate,
(70% solution in water) for synthesis
Expand
View Pricing
M4141
Methanesulfonic acid concentrate,
4 M (with 0.2% (w/v) tryptamine)
Expand
View Pricing