Dichloromethane solution
Synonym(s):
Methylene chloride solution, Dichloromethane solution
Empirical Formula (Hill Notation):
CH2Cl2
CAS Number:
Molecular Weight:
84.93
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique