Skip to Content
AU
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Acetic acid solution
Synonym(s):
Acetic acid solution
Linear Formula:
CH
3
CO
2
H
CAS Number:
64-19-7
Molecular Weight:
60.05
Products
Product Category
organic acids (1)
Brand
Sigma-Aldrich (1)
Supelco (4)
Formula Weight
Physical Form
Product Line
Agency
Quality Segment
Reaction Type
Technique
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
45754
Acetic acid solution,
suitable for HPLC
Expand
View Pricing
1.60305
Acetic acid solution,
1 M CH
3
COOH (1 N), Titripur
®
Expand
View Pricing
1.00062
Acetic acid solution,
for analysis EMSURE
®
Expand
View Pricing
1.59166
Acetic acid solution,
for analysis EMSURE
®
Reag. Ph Eur
Expand
View Pricing
1.60250
Acetic acid solution,
c(CH
3
COOH) = 0.1 mol/l (0.1 N), Titripur
®
Expand
View Pricing