Skip to Content
AU
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Lithium chloride solution
Linear Formula:
LiCl
CAS Number:
7447-41-8
Molecular Weight:
42.39
Products
Product Category
salts (1)
Brand
Millipore (1)
Sigma-Aldrich (1)
Color
Feature
Formula Weight
Physical Form
Purity
Quality Segment
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
L7026
Lithium chloride solution,
8 M, for molecular biology, ≥99%
Expand
View Pricing
5920-OP
Lithium chloride solution,
OmniPur
®
Grade, 8M
Expand
View Pricing