Sodium thiosulfate solution
Synonym(s):
Sodium thiosulfate solution
Linear Formula:
Na2S2O3
CAS Number:
Molecular Weight:
158.11
Products
Brand
Formula Weight
Physical Form
Product Line
Agency
Quality Segment
Application
Package Size
Reaction Type
Technique
Showing 1-6 of 6
Compare
Product Number
Description
Pricing