Proteomix® SAX-NP5 Strong Anion Exchange Column
Products
Product Category
Manufacturer
pH Value
Product Line
Agency
USP Suitability
Separation Technique
ID (mm)
Length (cm)
Package Size
Particle Size (µm)
Technique
Showing 1-8 of 8
Compare
Product Number
Description
Pricing