Skip to Content
AU
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Proteomix
®
HIC Phenyl-NP5, 5 μm, NP
Products
Product Category
columns (2)
Manufacturer
pH Value
Product Line
Separation Technique
ID (mm)
Length (mm)
Package Size
Particle Size (µm)
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
61876-U
Proteomix
®
HIC Phenyl-NP5, 5 μm, NP,
L × I.D. 100 mm × 4.6 mm
Expand
View Pricing
61874-U
Proteomix
®
HIC Phenyl-NP5, 5 μm, NP,
L × I.D. 35 mm × 4.6 mm
Expand
View Pricing
61878-U
Proteomix
®
HIC Phenyl-NP5, 5 μm, NP,
L × I.D. 50 mm × 7.8 mm
Expand
View Pricing