Nitinol-core (NIT) SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB)
Synonym(s):
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB)
Products
Brand
Sustainability
Feature
Length (cm)
Color
df (µm)
Size
Gauge
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing