Gliotoxin from Gliocladium fimbriatum
Empirical Formula (Hill Notation):
C13H14N2O4S2
CAS Number:
Molecular Weight:
326.39
Products
Product Category
Brand
Formula Weight
Activity Spectrum
Research Area
Quality Segment
Mode of Action
Showing 1-1 of 1
Compare
Product Number
Description
Pricing