Skip to Content
AU
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
α-Chymotrypsin from bovine pancreas
Synonym(s):
TLCK-Chymotrypsin
CAS Number:
9004-07-3
EC Number:
232-671-2
Products
Product Category
proteases (5)
Brand
Sigma-Aldrich (5)
Biological Source
Color
Physical Form
Tissue
Chemical Composition
Quality Segment
Application
Package Size
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
C4129
α-Chymotrypsin from bovine pancreas,
Type II, lyophilized powder, ≥40 units/mg protein
Expand
View Pricing
C3142
α-Chymotrypsin from bovine pancreas,
(TLCK treated to inactivate residual tryspin activity), Type VII, essentially salt-free, lyophilized powder, ≥40 units/mg protein
Expand
View Pricing
C7762
α-Chymotrypsin from bovine pancreas,
Type I-S, essentially salt-free, lyophilized powder
Expand
View Pricing
C6423
α-Chymotrypsin from bovine pancreas,
suitable for protein sequencing, salt-free, lyophilized powder
Expand
View Pricing
CHY5S
α-Chymotrypsin from bovine pancreas,
≥40 units/mg protein, vial of 5 mg
Expand
View Pricing