Skip to Content
Merck
tert-Butyl methyl ether suitable for HPLC, ≥99.8% (GC)

tert-Butyl methyl ether

tert-Butyl methyl ether suitable for HPLC, ≥99.8% (GC)
Synonym(s):
MTBE, Methyl tert-butyl ether, tert-Butyl methyl ether, tert-Butyl methyl ether
Linear Formula:
(CH3)3COCH3
CAS Number:
Molecular Weight:
88.15
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-18 of 18
Compare
Product Number
Description
Pricing