Diethyl ether
Synonym(s):
Ether, Ethyl ether, Diethyl ether
Linear Formula:
(CH3CH2)2O
CAS Number:
Molecular Weight:
74.12
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-17 of 17
Compare
Product Number
Description
Pricing