Skip to Content
AU
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
2-Methylbutane
Synonym(s):
Isopentane, 2-Methylbutane
Linear Formula:
CH
3
CH
2
CH(CH
3
)
2
CAS Number:
78-78-4
Molecular Weight:
72.15
EC Number:
201-142-8
Products
Product Category
solvents (6)
Brand
Sigma-Aldrich (5)
Supelco (1)
Boiling Point (°C)
Formula Weight
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Special Grade
Anhydrous (1)
Puriss (1)
Puriss P.A. (1)
Technique
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
M32631
2-Methylbutane,
ReagentPlus
®
, ≥99%
Expand
View Pricing
270342
2-Methylbutane,
suitable for HPLC, ≥99.5%
Expand
View Pricing
320404
2-Methylbutane,
ReagentPlus
®
, ≥99%
Expand
View Pricing
277258
2-Methylbutane,
anhydrous, ≥99%
Expand
View Pricing
1.06056
2-Methylbutane,
for spectroscopy Uvasol
®
Expand
View Pricing
59070
2-Methylbutane,
puriss. p.a., ≥99.5% (GC)
Expand
View Pricing