Skip to Content
Merck
1-Propanol natural, ≥98%, FG

1-Propanol

1-Propanol natural, ≥98%, FG
Synonym(s):
Propyl alcohol, 1-Propanol
Linear Formula:
CH3CH2CH2OH
CAS Number:
Molecular Weight:
60.10
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
Organoleptic Class
Reference Material Type
Special Grade
Technique