Skip to Content
Merck
Staurosporine from Streptomyces sp. ≥98% (HPLC), film

Staurosporine from Streptomyces sp.

Staurosporine from Streptomyces sp. ≥98% (HPLC), film
Synonym(s):
Antibiotic AM-2282
Empirical Formula (Hill Notation):
C28H26N4O3
CAS Number:
Molecular Weight:
466.53

Products

Product Category
Brand
Biological Source
Formula Weight
Physical Form
Purity
Activity Spectrum
Quality Segment
Mode of Action