Skip to Content
Merck
cis-Diamineplatinum(II) dichloride ≥99.9% trace metals basis

Cisplatin

cis-Diamineplatinum(II) dichloride ≥99.9% trace metals basis
Synonym(s):
Cisplatin, cis-Dichlorodiammine platinum(II), cis-Platinum(II) diammine dichloride, cis-Diammineplatinum(II) dichloride, cis-Platinum (65% Pt)
Linear Formula:
Pt(NH3)2Cl2
CAS Number:
Molecular Weight:
300.05

Products

Product Category
Brand
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Activity Spectrum
Quality Segment
Markush Class
Markush Group
Mode of Action
Reagent Type
Core