D-Pantothenic acid hemicalcium salt
Products
Product Category
Brand
Biological Source
Color
Feature
Formula Weight
Physical Form
Product Line
Agency
Quality Segment
Application
Optical Isomer
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing