Skip to Content
Merck
L-Ascorbic acid tested according to Ph. Eur.

L-Ascorbic acid

L-Ascorbic acid tested according to Ph. Eur.
Synonym(s):
L-Threoascorbic acid, Antiscorbutic factor, Vitamin C, L(+)-Ascorbic Acid, L-Ascorbic Acid, Free Acid
Empirical Formula (Hill Notation):
C6H8O6
CAS Number:
Molecular Weight:
176.12
EC Number:

Products

Product Category
Brand
Biological Source
Color
Feature
Formula Weight
Gene Alias
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Agency
Quality Segment
Markush Class
Markush Group
Optical Isomer
Technique