Skip to Content
Merck
TDA Reagent suitable for microbiology

Iron(III) chloride

TDA Reagent suitable for microbiology
Synonym(s):
Ferric chloride, Iron trichloride, Molysite, Iron(III) chloride, Iron(III) chloride solution, Ferric chloride solution
Linear Formula:
FeCl3
CAS Number:
Molecular Weight:
162.20

Products

Product Category
Brand
Sustainability
Feature
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Chemical Composition
Quality Segment
Application
Microorganism Suitability
Reagent Type
Core
Special Grade
Technique