Skip to Content
Merck
All Photos(1)

Key Documents

V800433

Sigma-Aldrich

Toluene

LR, sulfur free, 99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH3
CAS Number:
Molecular Weight:
92.14
Beilstein:
635760
EC Number:
MDL number:
UNSPSC Code:
12191501
PubChem Substance ID:
grade:
LR
bp:
110-111 °C (lit.)
vapor pressure:
22 mmHg ( 20 °C)
26 mmHg ( 25 °C)

grade

LR

vapor density

3.2 (vs air)

vapor pressure

22 mmHg ( 20 °C)
26 mmHg ( 25 °C)

product line

Vetec

Assay

99%

form

liquid

autoignition temp.

997 °F

expl. lim.

7 %

dilution

(for analytical testing)

color

colorless

refractive index

n/D 1.496 (lit.)

bp

110-111 °C (lit.)

mp

-93 °C (lit.)

density

0.865 g/mL at 25 °C (lit.)

SMILES string

Cc1ccccc1

InChI

1S/C7H8/c1-7-5-3-2-4-6-7/h2-6H,1H3

InChI key

YXFVVABEGXRONW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

39.9 °F - closed cup

Flash Point(C)

4.4 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ting Lei et al.
Small (Weinheim an der Bergstrasse, Germany), 11(24), 2946-2954 (2015-02-26)
Low-bandgap diketopyrrolopyrrole (DPP)-based polymers are used for the selective dispersion of semiconducting single-walled carbon nanotubes (s-SWCNTs). Through rational molecular design to tune the polymer-SWCNT interactions, highly selective dispersions of s-SWCNTs with diameters mainly around 1.5 nm are achieved. The influences
Marina Shalaeva et al.
Journal of medicinal chemistry, 56(12), 4870-4879 (2013-05-29)
This study demonstrates that ΔlogP(oct-tol) (difference between logP(octanol) and logP(toluene)) describes compounds propensity to form intramolecular hydrogen bonds (IMHB) and may be considered a privileged molecular descriptor for use in drug discovery and for prediction of IMHB in drug candidates.
Liang Guo et al.
Journal of chromatography. A, 1286, 9-15 (2013-03-19)
For the first time, a novel one-step sample preparation method that combines microwave assisted extraction and solvent bar microextraction (MAE-SBME) with analysis by gas chromatography-mass spectrometry (GC-MS), was developed for the fast and efficient determination of polycyclic aromatic hydrocarbons (PAHs)
Tayseer Mahdi et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 21(31), 11134-11142 (2015-06-26)
Frustrated Lewis pairs (FLPs) based on sterically encumbered anilines and the Lewis acid B(C6 F5 )3 were found to react with terminal alkynes effecting intermolecular hydroamination affording iminium alkynylborate species of the form [RPhNC(R')Me][R'CCB(C6 F5 )3 ]. In these cases
Christopher M Filley et al.
Journal of neuropathology and experimental neurology, 63(1), 1-12 (2004-01-30)
In recent decades the organic solvent toluene (methylbenzene) has emerged as one of the best-studied neurotoxins. Long-term and intense exposure to toluene vapors in humans who abuse spray paint and related substances has led to the recognition that toluene has

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service