Skip to Content
Merck
All Photos(1)

Key Documents

V800244

Sigma-Aldrich

Magnesium sulfate heptahydrate

LR, ≥99%

Synonym(s):

Epsom salts

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
MgSO4 · 7H2O
CAS Number:
Molecular Weight:
246.47
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
Assay:
≥99%
grade:
LR
form:
crystalline powder

grade

LR

vapor density

<0.01 (vs air)

vapor pressure

<0.1 mmHg ( 20 °C)

product line

Vetec

Assay

≥99%

form

crystalline powder

pH

5.0-8.2 (5% in water)

SMILES string

O.O.O.O.O.O.O.[Mg++].[O-]S([O-])(=O)=O

InChI

1S/Mg.H2O4S.7H2O/c;1-5(2,3)4;;;;;;;/h;(H2,1,2,3,4);7*1H2/q+2;;;;;;;;/p-2

InChI key

WRUGWIBCXHJTDG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ran D Goldman et al.
Pediatrics, 132(6), e1634-e1641 (2013-11-28)
Vaso-occlusive episodes (VOEs) are the most common complication of sickle cell disease in children. Treatment with magnesium seems to improve cellular hydration and may result in reduced vaso-occlusion. This study aimed to determine if intravenous (IV) magnesium sulfate (MgSO4) reduces
D H Salinger et al.
BJOG : an international journal of obstetrics and gynaecology, 120(7), 894-900 (2013-03-28)
To compare magnesium sulphate concentrations achieved by intramuscular and intravenous regimens used for the prevention of eclampsia. Low-resource obstetric hospitals in Nagpur and Vellore, India. Pregnant women at risk for eclampsia due to hypertensive disease. A pharmacokinetic study was performed
Karen J Gibbins et al.
Obstetrics and gynecology, 121(2 Pt 1), 235-240 (2013-01-25)
Clinical trials support the efficacy and safety of magnesium sulfate for cerebral palsy prevention. We evaluated the implementation of a clinical protocol for the use of magnesium for cerebral palsy prevention in our large women's hospital, focusing on uptake, indications
A P Morrison et al.
British journal of anaesthesia, 110(5), 702-712 (2013-03-28)
Spinal anaesthesia is the primary anaesthetic technique for many types of surgery. Adjuncts to the local anaesthetics (LA) used in spinal anaesthesia can exhibit undesirable side-effects, limiting their use, but magnesium may have advantages in this respect. We sought randomized
Chris Chin
Artificial organs, 37(1), 100-102 (2013-01-12)
Congenital heart disease (CHD) is relatively common and there are treatment options for virtually every form of CHD. With good care, the majority of babies can survive to adulthood; however, CHD remains a significant cause of death in infancy and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service