V800091
Bromobenzene
LR, ≥99%
Synonym(s):
1-Bromobenzene, Bromobenzol, Monobromobenzene, Phenyl bromide
About This Item
Recommended Products
grade
LR
Quality Level
vapor density
5.41 (vs air)
vapor pressure
10 mmHg ( 40 °C)
product line
Vetec™
Assay
≥99%
autoignition temp.
1051 °F
expl. lim.
36.5 %
refractive index
n20/D 1.559 (lit.)
bp
156 °C (lit.)
mp
−31 °C (lit.)
density
1.491 g/mL at 25 °C (lit.)
functional group
bromo
SMILES string
Brc1ccccc1
InChI
1S/C6H5Br/c7-6-4-2-1-3-5-6/h1-5H
InChI key
QARVLSVVCXYDNA-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 2 - Flam. Liq. 3 - Skin Irrit. 2
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Flash Point(F)
123.8 °F - closed cup
Flash Point(C)
51.0 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service