Key Documents
Z637602
BRAND® PD-Tip
maximum volume 100 μL, beta sterile/endotoxin-free
About This Item
UNSPSC Code:
41121600
NACRES:
NB.01
manufacturer/tradename:
BRAND 702404
maximum volume:
100 μL
sterility:
non-sterile
feature:
includes adapter: no
packaging:
pack of 100 ea (individually wrapped)
Agency
according to DIN 12 650
sterility
non-sterile
feature
includes adapter: no
packaging
pack of 100 ea (individually wrapped)
manufacturer/tradename
BRAND 702404
maximum volume
100 μL
General description
The new PLASTIBRAND PD-Tips with size encoding are the ideal system component for the HandyStep® electronic with automatic tip size recognition. The PD-Tips are DE-M marking and comply ISO 8655.
PLASTIBRAND PD-Tips fit with:
  • HandyStep® electronic, Gilson® Repetman, Rainin AutoRepTM E (automatic tip recognition), HandyStep, Rainin AutoRep M, Eppendorf® Multipette® 4780 and EDOS® 5221
  • PD-Tips are made from high-quality materials (cylinder: PP/piston: PE-HD, 0.1 mL: LCP)
  • An adapter is supplied standard for 25 mL and 50 mL
Features and Benefits
• Dispense with ease – even critical media• Save time with automatic size detection
Other Notes
BR702404 (India)
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
EDOS is a registered trademark of Eppendorf AG
Eppendorf is a registered trademark of Eppendorf AG
Gilson is a registered trademark of Gilson, Inc.
HandyStep is a registered trademark of BRAND GMBH + CO KG
Multipette is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)