Skip to Content
Merck
All Photos(2)

Key Documents

X1629

Sigma-Aldrich

Anti-XPG (C-terminal) antibody produced in rabbit

~1 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-COFS3, Anti-ERCC5, Anti-ERCM2, Anti-UVDR, Anti-XPGC

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

concentration

~1 mg/mL

technique(s)

indirect immunofluorescence: 2-4 μg/mL using HEK-293T cells expressing recombinant XPG fusion protein, fixed with paraformaldehyde-Triton
western blot: 1-2 μg/mL using whole cell lysates of COS7 cells expressing recombinant XPG fusion protein

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ERCC5(2073)

General description

Anti-XPG (C-terminal) is produced in rabbit using as immunogen a synthetic peptide corresponding to amino acids 1120-1137 of human XPG, conjugated to KLH via an N-terminal added cysteine residue. Xeroderma pigmentosum group G (XPG) is a DNA repair gene, that is mapped to human chromosome13q33. It is a member of the FEN-1 family of structure-specific nucleases and contains two highly conserved nuclease motifs known as the N and I regions separated by a large insertion. This gene spans around 69kb in length and has15 exons. It is highly polymorphic.

Immunogen

synthetic peptide corresponding to amino acids 1120-1137 of human XPG, conjugated to KLH via an N-terminal added cysteine residue.

Application

Anti-XPG (C-terminal) antibody produced in rabbit has been used in:
  • staining cells for xeroderma pigmentosum group G (XPG) detection
  • immunoblotting
  • immunofluorescence

Biochem/physiol Actions

Xeroderma pigmentosum group G (XPG) helps to maintain genomic stability. It also participates in RNA transcription by interacting with TFIIH, RNA polymerase II and Gadd45a. XPG also participates in nucleotide excision repair (NER) pathway. Mutations in the XPG gene cause the Cockayne syndrome which is characterized by severe growth defects, mental retardation and cachexia.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Domain swapping between FEN-1 and XPG defines regions in XPG that mediate nucleotide excision repair activity and substrate specificity
Hohl M, et al.
Nucleic Acids Research, 35(9), 3053-3063 (2007)
Lineage-specific control of TFIIH by MITF determines transcriptional homeostasis and DNA repair
Seoane M, et al.
Oncogene, 38(19), 3616-3616 (2019)
Loss of heterozygosity and lack of mutations of the XPG/ERCC5 DNA repair gene at 13q33 in prostate cancer
Hyytinen ER, et al.
Prostate, 41(3), 190-195 (1999)
XPG rs873601 G> A contributes to uterine leiomyoma susceptibility in a Southern Chinese population
Liu Z, et al.
Bioscience Reports, 38(5), BSR20181116-BSR20181116 (2018)
Identification of the XPG region that causes the onset of Cockayne syndrome by using Xpg mutant mice generated by the cDNA-mediated knock-in method
Shiomi N, et al.
Molecular and Cellular Biology, 24(9), 3712-3719 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service