SRP6283
Oxalate Decarboxylase
recombinant, expressed in HEK 293 cells, ≥98% (SDS-PAGE)
Synonym(s):
YvrK
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352202
NACRES:
NA.32
Recommended Products
recombinant
expressed in HEK 293 cells
Assay
≥98% (SDS-PAGE)
form
liquid
mol wt
45.9 kDa (384 aa + NT 6xHis-tag)
packaging
pkg of 1 mg
pkg of 100 μg
pkg of 20 μg
concentration
2 mg/mL
UniProt accession no.
shipped in
dry ice
storage temp.
−70°C
Gene Information
Bacillus subtilis ... OxdC(938620)
General description
N-methylation is one method by which drug and other xenobiotic compounds are metabolized by the liver. NNMT is an important cytosolic methyltransferase catalyzing the N-methylation of nicotinamide, pyridines and structural analogs, playing a crucial role in the biotransformation and detoxification of many xenobiotic compounds. In the N-methylation process, NNMT uses S-adenosyl methionine as the methyl donor and nicotinamide as methyl acceptor. NNMT is mostly expressed in the liver, and a lower expression is seen in the kidney, lung, skeletal muscle, placenta and heart. It may also play an important role in regulating biological processes related to N-methyl nicotinamide such as anti-inflammatory, anti-thrombotic, vasoprotective, and gastroprotective properties. Increased NNMT activity was reported in many kinds of tumors. NNMT is a potential biomarker and therapeutic target in caner diagnosis and treatment. NNMT serum levels have significance in the premature detection and in the management of patients with colorectal cancer. The recombinant protein includes hNNMT with N-terminal His-tag.
Physical form
2 mg/mL solution in HEPES Buffer containing 20% glycerol.
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service