Skip to Content
Merck
All Photos(1)

Key Documents

SRP3217

Sigma-Aldrich

LIGHT from mouse

recombinant, expressed in E. coli, ≥96% (SDS-PAGE), ≥96% (HPLC), suitable for cell culture

Synonym(s):

HVEM-L, TNFSF14

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

mouse

recombinant

expressed in E. coli

Assay

≥96% (HPLC)
≥96% (SDS-PAGE)

form

lyophilized

mol wt

20.1 kDa

packaging

pkg of 20 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

white to off-white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... TNFSF14(50930)

General description

LIGHT belongs to the TNF family of ligands, and can signal through the herpes virus entry mediator type A receptor (HVEM, TNFRSF14), LTbR, or bind to a decoy receptor, DcR3. It is expressed in splenocytes, activated PBL, CD+8 tumor infiltrating lymphocytes, granulocytes, and monocytes. LIGHT has the ability to active NFkB, to co-stimulate the activation of lymphocytes and to induce apoptosis in certain human tumor cells. Recombinant human LIGHT has a calculated mass of 19.3 kDa containing 177 amino acid residues. Recombinant murine LIGHT is a 20.1 kDa protein containing 183 amino acid residues.

Biochem/physiol Actions

LIGHT belongs to the TNF family of ligands, and can signal through the herpes virus entry mediator type A receptor (HVEM, TNFRSF14), LTbR, or bind to a decoy receptor, DcR3. Recombinant murine LIGHT is a 20.1 kDa protein containing 183 amino acid residues.

Sequence

MRLHQRLGDI VAHLPDGGKG SWEKLIQDQR SHQANPAAHL TGANASLIGI GGPLLWETRL GLAFLRGLTY HDGALVTMEP GYYYVYSKVQ LSGVGCPQGL ANGLPITHGL YKRTSRYPKE LELLVSRRSP CGRANSSRVW WDSSFLGGVV HLEAGEEVVV RVPGNRLVRP RDGTRSYFGA FMV

Physical form

Lyophilized from 20mM Tris, pH 8.5 + 100 mM Arginine.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rana Herro et al.
The Journal of allergy and clinical immunology, 136(3), 757-768 (2015-02-15)
Pulmonary fibrosis is characterized by excessive accumulation of collagen and α-smooth muscle actin in the lung. The key molecules that promote these phenotypes are of clinical interest. Thymic stromal lymphopoietin (TSLP) has been found at high levels in patients with
Rana Herro et al.
The Journal of investigative dermatology, 135(8), 2109-2118 (2015-03-20)
Several inflammatory diseases including scleroderma and atopic dermatitis display dermal thickening, epidermal hypertrophy, or excessive accumulation of collagen. Factors that might promote these features are of interest for clinical therapy. We previously reported that LIGHT, a TNF superfamily molecule, mediated
J A Harrop et al.
The Journal of biological chemistry, 273(42), 27548-27556 (1998-10-09)
Herpesvirus entry mediator (HVEM), a member of the tumor necrosis factor (TNF) receptor family, mediates herpesvirus entry into cells during infection. Upon overexpression, HVEM activates NF-kappaB and AP-1 through a TNF receptor-associated factor (TRAF)-mediated mechanism. Using an HVEM-Fc fusion protein
D N Mauri et al.
Immunity, 8(1), 21-30 (1998-02-14)
Herpes simplex virus (HSV) 1 and 2 infect activated T lymphocytes by attachment of the HSV envelope glycoprotein D (gD) to the cellular herpesvirus entry mediator (HVEM), an orphan member of the tumor necrosis factor receptor superfamily. Here, we demonstrate
Kari Otterdal et al.
Clinical and translational gastroenterology, 6, e95-e95 (2015-07-03)
The tumor necrosis factor superfamily member 14, LIGHT (homologous to lymphotoxin, exhibits inducible expression, and competes with HSV glycoprotein D for herpes virus entry mediator (HVEM), a receptor expressed by T lymphocytes), has been involved in various autoimmune disorders and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service