SML2547
KCC-07
≥98% (HPLC)
Synonym(s):
3-[[4-(2-Pyridinyl)-2-thiazolyl]amino]phenol
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C14H11N3OS
CAS Number:
Molecular Weight:
269.32
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Recommended Products
Assay
≥98% (HPLC)
form
powder
color
white to beige
solubility
DMSO: 2 mg/mL, clear
storage temp.
2-8°C
SMILES string
OC1=CC(NC2=NC(C3=NC=CC=C3)=CS2)=CC=C1
InChI key
GIGNWEDIMLUWCT-UHFFFAOYSA-N
Biochem/physiol Actions
KCC-07 is a brain penetrant, potent and selective inhibitor of MBD2 (methyl-CpG-binding domain protein 2) that inhibits binding of MBD2 to methylated DNA and activates brain specific angiogenesis inhibitor 1 (BAI1) inducing anti-proliferative BAI1/p53/p21 signaling. KCC-07 inhibits medulloblastoma cell proliferation, and inhibits tumor growth in mice xenograft models of medulloblastoma.
brain penetrant, potent and selective inhibitor of MBD2 that inducing anti-proliferative BAI1/p53/p21 signaling
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Dan Zhu et al.
Cancer cell, 33(6), 1004-1016 (2018-06-13)
Adhesion G protein-coupled receptors (ADGRs) encompass 33 human transmembrane proteins with long N termini involved in cell-cell and cell-matrix interactions. We show the ADGRB1 gene, which encodes Brain-specific angiogenesis inhibitor 1 (BAI1), is epigenetically silenced in medulloblastomas (MBs) through a methyl-CpG
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service