Skip to Content
Merck
All Photos(2)

Key Documents

SAB5600106

Sigma-Aldrich

Anti-Phospho-AMPA Receptor antibody, GluR 1 subunit (Ser845), Rabbit Monoclonal

recombinant, expressed in HEK 293 cells, clone RM296, purified immunoglobulin

Synonym(s):

Glutamate Receptor 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
clone:
RM296, monoclonal
recombinant monoclonal
application:
IHC
WB
species reactivity:
human
technique(s):
immunohistochemistry: 1:100-1:200
western blot: 1:100-1:200
citations:
3

recombinant

expressed in HEK 293 cells

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

RM296, monoclonal
recombinant monoclonal

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:100-1:200
western blot: 1:100-1:200

isotype

IgG

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pSer845)

Gene Information

human ... GRIA1(2890)

General description

GLUR1/GRIA1 (glutamate ionotropic receptor AMPA type subunit 1) is located on human chromosome 5q33.2. It is expressed mainly in brain areas.

Specificity

This antibody reacts to human Glutamate Receptor 1, AMPA Receptor (GluR1) only when phosphorylated at Ser845. There is no cross-reactivity to GluR1 that is not phosphorylated at Ser845. This antibody may also react to mouse or rat Phospho-AMPA Receptor (GluR1) (Ser845) as predicted by immunogen homology.

Immunogen

A phospho-peptide corresponding to human Phospho-Glutamate Receptor 1 (GluR1) (Ser845).

Biochem/physiol Actions

GLUR1/GRIA1 (glutamate ionotropic receptor AMPA type subunit 1) plays an important role in long-term potentiation (LTP). It is associated with migraine.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline containing 50% glycerol, 1% BSA and 0.09% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Common variants in the regulative regions of GRIA1 and GRIA3 receptor genes are associated with migraine susceptibility
Formicola D, et al.
BMC Medical Genetics, 11(1), 103-103 (2010)
Functional organization of the GluR1 glutamate receptor promoter
Borges K and Dingledine R
The Journal of Biological Chemistry, 276(28), 25929-25938 (2001)
Selective disruption of stimulus-reward learning in glutamate receptor gria1 knock-out mice
Mead AN and Stephens DN
The Journal of Neuroscience, 23(3), 1041-1048 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service