Skip to Content
Merck
All Photos(1)

Key Documents

SAB2501598

Sigma-Aldrich

Anti-CD244 antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-2B4, Anti-NAIL, Anti-NKR2B4, CD244 molecule, natural killer cell receptor 2B4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunoprecipitation (IP): suitable
indirect ELISA: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CD244(51744)

Immunogen

Peptide with sequence EFLTIpYEDVKD, from the internal region of the protein sequence according to NP_057466.1

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.5 mg/mL in 20mM Tris (pH 7.3) and 150mM NaCl with 0.02% sodium azide and 0.5% bovine serum albumin.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yoshihiro Ohue et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 20(19), 5052-5063 (2014-08-16)
The cancer/testis antigen XAGE1 (GAGED2a) is expressed in approximately 40% of advanced lung adenocarcinomas. We investigated the clinical relevance of the XAGE1 (GAGED2a) immune responses in patients with advanced lung adenocarcinoma. The XAGE1 (GAGED2a) antigen expression and EGFR mutation were
Natasja Nielsen et al.
Immunology, 142(4), 581-593 (2014-03-29)
Rheumatoid arthritis (RA) is an autoimmune disease characterized by chronic inflammation and synovial hyperplasia leading to progressive joint destruction. Fibroblast-like synoviocytes (FLS) are central components of the aggressive, tumour-like synovial structure termed pannus, which invades the joint space and cartilage.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service