Skip to Content
Merck
All Photos(2)

Key Documents

SAB2104365

Sigma-Aldrich

Anti-FOXC1, (N-terminal) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-ARA, Anti-FKHL7, Anti-FREAC3, Anti-IGDA, Anti-IHG1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

57 kDa

species reactivity

rat, human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FOXC1(2296)

Immunogen

Synthetic peptide directed towards the N terminal region of human FOXC1

Biochem/physiol Actions

FOXC1 belongs to the forkhead family of transcription factors which is characterized by a distinct DNA-binding forkhead domain. The specific function of this gene has not yet been determined; however, it has been shown to play a role in the regulation of

Sequence

Synthetic peptide located within the following region: GGYTAMPAPMSVYSHPAHAEQYPGGMARAYGPYTPQPQPKDMVKPPYSYI

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E Orta-Salazar et al.
Life sciences, 116(2), 83-89 (2014-09-28)
The cholinergic system is one of the neurotransmitter systems altered in Alzheimer's disease (AD), the most common form of human dementia. The objective of this work was to determine the REST/NRSF involvement in altered ChAT expression in the neocortex and
Irene E Whitney et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 34(30), 10109-10121 (2014-07-25)
The retina contains two populations of cholinergic amacrine cells, one positioned in the ganglion cell layer (GCL) and the other in the inner nuclear layer (INL), that together comprise ∼1/2 of a percent of all retinal neurons. The present study

Global Trade Item Number

SKUGTIN
SAB2104365-50UG
SAB2104365-100UL4061836177447

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service