Skip to Content
Merck
All Photos(3)

Key Documents

SAB1409297

Sigma-Aldrich

Monoclonal Anti-SMAD7 antibody produced in mouse

clone 3G8, purified immunoglobulin, buffered aqueous solution

Synonym(s):

CRCS3, FLJ16482, MADH7, MADH8

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3G8, monoclonal

form

buffered aqueous solution

mol wt

antigen 36.85 kDa

species reactivity

human

technique(s)

indirect ELISA: suitable
indirect immunofluorescence: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SMAD7(4092)

Immunogen

SMAD7 (NP_005895, 160 a.a. ~ 260 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
CKVFRWPDLRHSSEVKRLCCCESYGKINPELVCCNPHHLSRLCELESPPPPYSRYPMDFLKPTADCPDAVPSSAETGGTNYLAPGGLSDSQLLLEPGDRSH

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Busheng Zhang et al.
PloS one, 9(6), e100298-e100298 (2014-06-19)
MicroRNAs (miRNAs) regulate a lot of physiological and pathological processes, including myocardial ischemia/reperfusion. Recent studies reported that knockdown of miR-92a could attenuate ischemia/reperfusion-induced myocardial injury. In the present study, we examined the potential anti-apoptotic effects of miR-92a in a rat
Yunchuan Li et al.
Chinese medical journal, 127(19), 3449-3453 (2014-10-02)
Aberrant epithelial repair has been observed in chronic rhinosinusitis (CRS) patients; however, the mechanism of epithelial cell repair regulation is unclear. Epidermal growth factor (EGF) plays an important role in regulating epithelial cell repair in lower airway and may be

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service